siRNA Details
Gene Symbol: Usp42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105909.1 | NP_001099379.1 | 12 | 1532 | ||
XM_008768948.1 | XP_008767170.1 | 12 | 1542 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Usp42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105909.1 | NP_001099379.1 | 12 | 1532 | ||
XM_008768948.1 | XP_008767170.1 | 12 | 1542 |