siRNA Details
Gene Symbol: Wipi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007615.1 | NP_001007616.1 | 11 | 1288 | ||
GenBank mRNA | BC079184.1 | - | 11 | 1288 | 7131054 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wipi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007615.1 | NP_001007616.1 | 11 | 1288 | ||
GenBank mRNA | BC079184.1 | - | 11 | 1288 | 7131054 |