siRNA Details
| Gene Symbol: Wipi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007615.1 | NP_001007616.1 | 4 | 574 | ||
| GenBank mRNA | BC079184.1 | - | 4 | 574 | 7131054 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wipi2| Gene Symbol: Wipi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007615.1 | NP_001007616.1 | 4 | 574 | ||
| GenBank mRNA | BC079184.1 | - | 4 | 574 | 7131054 | |