siRNA Details
| Gene Symbol: Dhrsx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105914.1 | NP_001099384.1 | 4 | 328 | ||
| XM_006248993.2 | XP_006249055.1 | 4 | 379 | |||
| XM_006248995.2 | XP_006249057.1 | 4 | 380 | |||
| XM_006248997.1 | XP_006249059.1 | 4 | 623 | |||
| GenBank mRNA | BC167053.1 | - | 4 | 307 | 7110020 | |

Dhrsx
Genomic Map