siRNA Details
| Gene Symbol: RGD1560281 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769099.1 | XP_008767321.1 | 3 | 209 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1560281| Gene Symbol: RGD1560281 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769099.1 | XP_008767321.1 | 3 | 209 | ||