siRNA Details
| Gene Symbol: Tfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105916.1 | NP_001099386.1 | 18 | 1985 | ||
| XM_006249116.2 | XP_006249178.1 | 17 | 1937 | |||
| XM_006249117.1 | XP_006249179.1 | 18 | 1953 | |||
| XM_006249118.1 | XP_006249180.1 | 18 | 1956 | |||
| XM_008769125.1 | XP_008767347.1 | 17 | 2066 | |||
| XM_008769126.1 | XP_008767348.1 | 17 | 1896 | |||
| XM_008769128.1 | XP_008767350.1 | 17 | 1937 | |||
| GenBank mRNA | BC166451.1 | - | 17 | 2050 | 8371649 | |

Tfr2
Genomic Map