siRNA Details
| Gene Symbol: Plod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178101.3 | NP_835202.2 | 5 | 870 | ||
| GenBank mRNA | AJ430859.1 | - | 5 | 609 | ||
| BC085683.1 | - | 5 | 650 | 7110561 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plod3| Gene Symbol: Plod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178101.3 | NP_835202.2 | 5 | 870 | ||
| GenBank mRNA | AJ430859.1 | - | 5 | 609 | ||
| BC085683.1 | - | 5 | 650 | 7110561 | ||