siRNA Details
Gene Symbol: Ccl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013045.1 | NP_001013063.1 | 2 | 178 | ||
GenBank mRNA | BC087576.1 | - | 2 | 178 | 7307568 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013045.1 | NP_001013063.1 | 2 | 178 | ||
GenBank mRNA | BC087576.1 | - | 2 | 178 | 7307568 |