siRNA Details
Gene Symbol: Wbscr28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249213.2 | XP_006249275.1 | 2 | 897 | ||
XM_213758.8 | XP_213758.4 | 2 | 256 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wbscr28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249213.2 | XP_006249275.1 | 2 | 897 | ||
XM_213758.8 | XP_213758.4 | 2 | 256 |