siRNA Details
| Gene Symbol: Sbds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008289.1 | NP_001008290.1 | 2 | 203 | ||
| GenBank mRNA | BC086335.1 | - | 2 | 203 | 7301062 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sbds| Gene Symbol: Sbds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008289.1 | NP_001008290.1 | 2 | 203 | ||
| GenBank mRNA | BC086335.1 | - | 2 | 203 | 7301062 | |