siRNA Details
| Gene Symbol: Tmem248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004204.2 | NP_001004204.1 | 9 | 1384 | ||
| XM_006249230.2 | XP_006249292.1 | 6 | 1086 | |||
| XM_006249231.2 | XP_006249293.1 | 6 | 1007 | |||
| GenBank mRNA | BC079161.1 | - | 6 | 1058 | 7129411 | |
| BC090318.1 | - | 9 | 1384 | 7113430 | ||

Tmem248
Genomic Map