siRNA Details
| Gene Symbol: Tpst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011903.2 | NP_001011903.2 | 3 | 1163 | ||
| XM_006249234.1 | XP_006249296.1 | 3 | 1167 | |||
| XM_008769133.1 | XP_008767355.1 | 4 | 1177 | |||
| GenBank mRNA | BC078928.1 | - | 4 | 1101 | 7106945 | |
| BC105826.1 | - | 3 | 1163 | 7381985 | ||

Tpst1
Genomic Map