siRNA Details
| Gene Symbol: Cct6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033684.1 | NP_001028856.1 | 3 | 244 | ||
| GenBank mRNA | BC104703.1 | - | 3 | 244 | 7300674 | |
| FQ216833.1 | - | 3 | 262 | |||
| FQ218432.1 | - | 3 | 291 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cct6a| Gene Symbol: Cct6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033684.1 | NP_001028856.1 | 3 | 244 | ||
| GenBank mRNA | BC104703.1 | - | 3 | 244 | 7300674 | |
| FQ216833.1 | - | 3 | 262 | |||
| FQ218432.1 | - | 3 | 291 | |||