siRNA Details
| Gene Symbol: Mmp17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105925.1 | NP_001099395.1 | 9 | 1354 | ||
| XM_008769195.1 | XP_008767417.1 | 9 | 2637 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmp17| Gene Symbol: Mmp17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105925.1 | NP_001099395.1 | 9 | 1354 | ||
| XM_008769195.1 | XP_008767417.1 | 9 | 2637 | |||