siRNA Details
| Gene Symbol: Gtf2h3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024236.1 | NP_001019407.1 | 11 | 741 | ||
| GenBank mRNA | BC093380.1 | - | 11 | 741 | 7388486 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gtf2h3| Gene Symbol: Gtf2h3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024236.1 | NP_001019407.1 | 11 | 741 | ||
| GenBank mRNA | BC093380.1 | - | 11 | 741 | 7388486 | |