siRNA Details
Gene Symbol: Rilpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004205.1 | NP_001004205.1 | 1 | 450 | ||
GenBank mRNA | BC079132.1 | - | 1 | 450 | 7127390 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rilpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004205.1 | NP_001004205.1 | 1 | 450 | ||
GenBank mRNA | BC079132.1 | - | 1 | 450 | 7127390 |