siRNA Details
| Gene Symbol: Ogfod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105927.1 | NP_001099397.1 | 6 | 673 | ||
| XM_006249331.1 | XP_006249393.1 | 6 | 673 | |||
| GenBank mRNA | BC167103.1 | - | 4 | 834 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ogfod2| Gene Symbol: Ogfod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105927.1 | NP_001099397.1 | 6 | 673 | ||
| XM_006249331.1 | XP_006249393.1 | 6 | 673 | |||
| GenBank mRNA | BC167103.1 | - | 4 | 834 | ||