siRNA Details
| Gene Symbol: Atxn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769281.1 | XP_008767503.1 | 14, 15 | 5213 | ||
| XM_008769282.1 | XP_008767504.1 | 14, 15 | 5213 | |||
| XM_008769283.1 | XP_008767505.1 | 14, 15 | 5212 | |||
| XM_008769284.1 | XP_008767506.1 | 14, 15 | 5212 | |||
| XM_008769285.1 | XP_008767507.1 | 14, 15 | 5212 | |||
| XM_008769286.1 | XP_008767508.1 | 14, 15 | 5212 | |||
| XM_008769287.1 | XP_008767509.1 | 14, 15 | 5213 | |||
| XM_008769288.1 | XP_008767510.1 | 14, 15 | 5212 | |||
| XM_008769289.1 | XP_008767511.1 | 14, 15 | 5213 | |||
| XM_008769290.1 | XP_008767512.1 | 14, 15 | 5212 | |||
| XM_008769292.1 | XP_008767514.1 | 13, 14 | 5003 | |||
| XM_008769293.1 | XP_008767515.1 | 14, 15 | 5212 | |||
| XM_008769294.1 | XP_008767516.1 | 14, 15 | 5213 | |||
| XM_008769295.1 | XP_008767517.1 | 14, 15 | 5213 | |||
| GenBank mRNA | BC168999.1 | - | 11, 12 | 1706 | ||

Atxn2
Genomic Map