siRNA Details
| Gene Symbol: Vps29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105932.1 | NP_001099402.1 | 4 | 589 | ||
| XM_006249362.2 | XP_006249424.1 | 5 | 615 | |||
| GenBank mRNA | AB578912.1 | - | 3 | 339 | ||
| BC167055.1 | - | 5 | 576 | 7302374 | ||
| FQ224287.1 | - | 5 | 597 | |||

Vps29
Genomic Map