siRNA Details
| Gene Symbol: APCS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001639.3 | NP_001630.1 | 2 | 484 | ||
| GenBank mRNA | BC007039.1 | - | 2 | 484 | 3935086 | |
| BC007058.1 | - | 2 | 484 | 3934872 | ||
| BC070178.1 | - | 2 | 158 | |||
| BT006750.1 | - | 2 | 388 | |||
| CR450313.1 | - | 2 | 388 | |||
| FJ460512.1 | - | 2 | 484 | |||
| M10944.1 | - | 2 | 388 | |||
| X04608.1 | - | 2 | 484 | |||

APCS
Genomic Map