siRNA Details
| Gene Symbol: Srsf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009255.1 | NP_001009255.1 | 3 | 545 | ||
| GenBank mRNA | BC087684.1 | - | 3 | 545 | 7309111 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srsf9| Gene Symbol: Srsf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009255.1 | NP_001009255.1 | 3 | 545 | ||
| GenBank mRNA | BC087684.1 | - | 3 | 545 | 7309111 | |