siRNA Details
| Gene Symbol: Tfip11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008291.1 | NP_001008292.1 | 14 | 2695 | ||
| GenBank mRNA | BC085809.1 | - | 14 | 2695 | 7113767 | |
| DQ342031.1 | - | 14 | 2694 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tfip11| Gene Symbol: Tfip11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008291.1 | NP_001008292.1 | 14 | 2695 | ||
| GenBank mRNA | BC085809.1 | - | 14 | 2695 | 7113767 | |
| DQ342031.1 | - | 14 | 2694 | |||