siRNA Details
| Gene Symbol: Pgam5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025272.1 | NP_001020443.1 | 4, 5 | 617 | ||
| GenBank mRNA | BC092607.1 | - | 4, 5 | 617 | 7309810 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pgam5| Gene Symbol: Pgam5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025272.1 | NP_001020443.1 | 4, 5 | 617 | ||
| GenBank mRNA | BC092607.1 | - | 4, 5 | 617 | 7309810 | |