siRNA Details
| Gene Symbol: Foxn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105935.1 | NP_001099405.1 | 10 | 1230 | ||
| XM_006249493.2 | XP_006249555.1 | 10 | 1771 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxn4| Gene Symbol: Foxn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105935.1 | NP_001099405.1 | 10 | 1230 | ||
| XM_006249493.2 | XP_006249555.1 | 10 | 1771 | |||