siRNA Details
| Gene Symbol: Ficd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010946.1 | NP_001010946.1 | 2 | 886 | ||
| XM_008769320.1 | XP_008767542.1 | 3 | 783 | |||
| XM_008769321.1 | XP_008767543.1 | 3 | 713 | |||
| GenBank mRNA | BC079197.1 | - | 2 | 886 | 7131463 | |

Ficd
Genomic Map