siRNA Details
| Gene Symbol: Sgsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105937.2 | NP_001099407.2 | 8 | 760 | ||
| XM_008769322.1 | XP_008767544.1 | 8 | 1099 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sgsm1| Gene Symbol: Sgsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105937.2 | NP_001099407.2 | 8 | 760 | ||
| XM_008769322.1 | XP_008767544.1 | 8 | 1099 | |||