siRNA Details
| Gene Symbol: Tmem132d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198727.1 | NP_942022.1 | 9 | 3245 | ||
| XM_006249266.2 | XP_006249328.2 | 9 | 3871 | |||
| GenBank mRNA | AB100356.1 | - | 9 | 3245 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem132d| Gene Symbol: Tmem132d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198727.1 | NP_942022.1 | 9 | 3245 | ||
| XM_006249266.2 | XP_006249328.2 | 9 | 3871 | |||
| GenBank mRNA | AB100356.1 | - | 9 | 3245 | ||