siRNA Details
| Gene Symbol: B3gnt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105938.1 | NP_001099408.1 | 1 | 1016 | ||
| XM_006249335.2 | XP_006249397.1 | 2 | 1207 | |||
| XM_006249336.2 | XP_006249398.1 | 2 | 983 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B3gnt4| Gene Symbol: B3gnt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105938.1 | NP_001099408.1 | 1 | 1016 | ||
| XM_006249335.2 | XP_006249397.1 | 2 | 1207 | |||
| XM_006249336.2 | XP_006249398.1 | 2 | 983 | |||