siRNA Details
| Gene Symbol: Diablo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008292.1 | NP_001008293.1 | 2, 3 | 190 | ||
| GenBank mRNA | BC086366.1 | - | 2, 3 | 190 | 7305418 | |
| FQ219826.1 | - | 2, 3 | 183 | |||
| Gene Symbol: LOC100360940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729540.3 | XP_002729586.2 | 1, 2 | 220 | ||

Diablo
Genomic Map