siRNA Details
| Gene Symbol: Olr875 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000054.1 | NP_001000054.1 | 1 | 867 | ||
| Gene Symbol: Olr876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000708.1 | NP_001000708.1 | 1 | 867 | ||

Olr875
Genomic Map