siRNA Details
| Gene Symbol: Olr907 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001357.1 | NP_001001357.1 | 1 | 142 | ||
| Gene Symbol: Olr917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001354.1 | NP_001001354.1 | 1 | 142 | ||

Olr907
Genomic Map