siRNA Details
| Gene Symbol: Olr883 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001358.1 | NP_001001358.1 | 2 | 142 | ||
| Gene Symbol: Olr910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001359.1 | NP_001001359.1 | 2 | 142 | ||

Olr883
Genomic Map