siRNA Details
| Gene Symbol: Olr1061 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000065.1 | NP_001000065.1 | 1 | 233 | ||
| Gene Symbol: Olr927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000588.1 | NP_001000588.1 | 1 | 233 | ||

Olr1061
Genomic Map