siRNA Details
| Gene Symbol: Olr1029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001366.1 | NP_001001366.1 | 1 | 238 | ||
| Gene Symbol: Olr1002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001371.1 | NP_001001371.1 | 1 | 238 | ||
| Gene Symbol: Olr956 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001381.1 | NP_001001381.1 | 1 | 238 | ||

Olr1029
Genomic Map