siRNA Details
| Gene Symbol: Asna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100505.1 | NP_001093975.1 | 1 | 59 | ||
| XM_008772367.1 | XP_008770589.1 | 2 | 880 | |||
| XM_008772368.1 | XP_008770590.1 | 2 | 1008 | |||
| XM_008772369.1 | XP_008770591.1 | 2 | 1003 | |||
| GenBank mRNA | CB700861.1 | - | 1 | 59 | ||

Asna1
Genomic Map