siRNA Details
| Gene Symbol: Fbxw9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081634.1 | NP_001075103.1 | 5 | 915 | ||
| GenBank mRNA | BC084706.1 | - | 4 | 958 | ||
| BC166590.1 | - | 4 | 932 | 7938435 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxw9| Gene Symbol: Fbxw9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081634.1 | NP_001075103.1 | 5 | 915 | ||
| GenBank mRNA | BC084706.1 | - | 4 | 958 | ||
| BC166590.1 | - | 4 | 932 | 7938435 | ||