siRNA Details
| Gene Symbol: Serpinb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105948.1 | NP_001099418.1 | 2 | 340 | ||
| XM_008769428.1 | XP_008767650.1 | 2 | 602 | |||
| XM_008769429.1 | XP_008767651.1 | 3 | 1174 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpinb8| Gene Symbol: Serpinb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105948.1 | NP_001099418.1 | 2 | 340 | ||
| XM_008769428.1 | XP_008767650.1 | 2 | 602 | |||
| XM_008769429.1 | XP_008767651.1 | 3 | 1174 | |||