siRNA Details
| Gene Symbol: Cdk18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100506.1 | NP_001093976.1 | 4 | 361 | ||
| XM_006249750.2 | XP_006249812.1 | 5 | 448 | |||
| XM_006249751.2 | XP_006249813.1 | 5 | 509 | |||
| XM_006249752.2 | XP_006249814.1 | 5 | 969 | |||
| XM_006249753.2 | XP_006249815.1 | 5 | 482 | |||
| XM_008769441.1 | XP_008767663.1 | 5 | 787 | |||
| GenBank mRNA | AB005541.1 | - | 5 | 441 | ||
| BC082045.1 | - | 4 | 361 | 7131722 | ||

Cdk18
Genomic Map