siRNA Details
| Gene Symbol: Pik3c2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105951.1 | NP_001099421.1 | 27, 28 | 4721 | ||
| XM_006249754.2 | XP_006249816.1 | 28, 29 | 4824 | |||
| XM_006249756.2 | XP_006249818.1 | 27, 28 | 4267 | |||
| XM_008769442.1 | XP_008767664.1 | 28, 29 | 5664 | |||

Pik3c2b
Genomic Map