siRNA Details
| Gene Symbol: Chit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079689.2 | NP_001073157.1 | 7 | 1171 | ||
| NM_001270846.1 | NP_001257775.1 | 8 | 820 | |||
| NM_001270847.1 | NP_001257776.1 | 7 | 719 | |||
| NM_001270848.1 | NP_001257777.1 | 7 | 804 | |||
| XM_008769542.1 | XP_008767764.1 | 6 | 1100 | |||
| GenBank mRNA | DQ286232.1 | - | 6 | 697 | ||
| DQ923316.1 | - | 6 | 642 | |||
| DQ923317.1 | - | 7 | 804 | |||
| DQ923318.1 | - | 7 | 720 | |||
| DQ923319.1 | - | 8 | 820 | |||
| DQ923320.1 | - | 7 | 1162 | |||

Chit1
Genomic Map