siRNA Details
| Gene Symbol: RGD1308112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249895.2 | XP_006249957.1 | 5 | 1539 | ||
| XM_008769575.1 | XP_008767797.1 | 4 | 1456 | |||
| XM_008769576.1 | XP_008767798.1 | 5 | 1534 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1308112| Gene Symbol: RGD1308112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249895.2 | XP_006249957.1 | 5 | 1539 | ||
| XM_008769575.1 | XP_008767797.1 | 4 | 1456 | |||
| XM_008769576.1 | XP_008767798.1 | 5 | 1534 | |||