siRNA Details
| Gene Symbol: Zbtb41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191670.1 | NP_001178599.1 | 1 | 626 | ||
| XM_006249937.2 | XP_006249999.1 | 2 | 855 | |||
| XM_006249938.2 | XP_006250000.1 | 2 | 820 | |||
| XM_006249939.2 | XP_006250001.1 | 2 | 846 | |||
| XM_006249940.2 | XP_006250002.1 | 2 | 898 | |||

Zbtb41
Genomic Map