siRNA Details
| Gene Symbol: Zbtb41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191670.1 | NP_001178599.1 | 1 | 350 | ||
| XM_006249937.2 | XP_006249999.1 | 2 | 579 | |||
| XM_006249938.2 | XP_006250000.1 | 2 | 544 | |||
| XM_006249939.2 | XP_006250001.1 | 2 | 570 | |||
| XM_006249940.2 | XP_006250002.1 | 2 | 622 | |||
| GenBank mRNA | FQ218188.1 | - | 2 | 506 | ||

Zbtb41
Genomic Map