siRNA Details
| Gene Symbol: F13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105956.1 | NP_001099426.1 | 9 | 1445 | ||
| XM_008769547.1 | XP_008767769.1 | 10 | 1696 | |||
| GenBank mRNA | BC160876.1 | - | 10 | 1629 | 7369453 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

F13b| Gene Symbol: F13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105956.1 | NP_001099426.1 | 9 | 1445 | ||
| XM_008769547.1 | XP_008767769.1 | 10 | 1696 | |||
| GenBank mRNA | BC160876.1 | - | 10 | 1629 | 7369453 | |