siRNA Details
Gene Symbol: Rgs18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047084.1 | NP_001040549.1 | 5 | 560 | ||
XM_008769548.1 | XP_008767770.1 | 5 | 586 | |||
GenBank mRNA | AY651776.1 | - | 5 | 560 |
