siRNA Details
| Gene Symbol: Rgs18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047084.1 | NP_001040549.1 | 2 | 142 | ||
| XM_008769548.1 | XP_008767770.1 | 2 | 168 | |||
| GenBank mRNA | AY651776.1 | - | 2 | 142 | ||
| Gene Symbol: LOC103690097 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769583.1 | XP_008767805.1 | 391 | |||

Rgs18
Genomic Map