siRNA Details
| Gene Symbol: Swt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105960.1 | NP_001099430.1 | 18 | 2708 | ||
| XM_008769613.1 | XP_008767835.1 | 17 | 2548 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Swt1| Gene Symbol: Swt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105960.1 | NP_001099430.1 | 18 | 2708 | ||
| XM_008769613.1 | XP_008767835.1 | 17 | 2548 | |||