siRNA Details
| Gene Symbol: Shcbp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105961.2 | NP_001099431.1 | 5 | 920 | ||
| GenBank mRNA | BC160910.1 | - | 5 | 920 | 7456157 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shcbp1l| Gene Symbol: Shcbp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105961.2 | NP_001099431.1 | 5 | 920 | ||
| GenBank mRNA | BC160910.1 | - | 5 | 920 | 7456157 | |