siRNA Details
| Gene Symbol: Cenpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033061.2 | NP_001028233.2 | 5 | 1343 | ||
| NM_001244761.1 | NP_001231690.1 | 4 | 1257 | |||
| XM_006250099.2 | XP_006250161.1 | 5 | 1395 | |||
| XM_006250100.2 | XP_006250162.1 | 4 | 1308 | |||
| GenBank mRNA | BC079467.1 | - | 5 | 1343 | 7122576 | |
| BC089893.1 | - | 4 | 1257 | |||
| BC103642.1 | - | 5 | 1336 | 7300924 | ||

Cenpl
Genomic Map