siRNA Details
| Gene Symbol: AIRE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000383.3 | NP_000374.1 | 14 | 1977 | ||
| GenBank mRNA | AB006682.1 | - | 14 | 1935 | ||
| AB006683.1 | - | 8 | 1453 | |||
| AB006685.1 | - | 9 | 1371 | |||
| Z97990.1 | - | 14 | 1928 | |||

AIRE
Genomic Map